Gene Transcripts
- Gene Symbol
- Ppp1r12b
- Entrez Gene ID
- 329251
- Gene Name
- protein phosphatase 1, regulatory (inhibitor) subunit 12B
- Gene Aliases
- 1810037O03Rik, 9530009M10Rik, AI132431, Mypt2
- Chromosome Location
- Chr. 1: 134754665 - 134955965 on Build GRCm38
- UniGene ID
- Mm.188709
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015606.3 |
|
2 |
2989 |
|
GenBank mRNA |
AK035285.1 |
- |
2 |
3120 |
|
AK051953.1 |
- |
2 |
2867 |
|
View Full Product Details