Gene Transcripts
- Gene Symbol
- FAM181A-AS1
- Entrez Gene ID
- 283592
- Gene Name
- FAM181A antisense RNA 1
- Gene Aliases
- C14orf86
- Chromosome Location
- Chr. 14: 93904730 - 93926372 on Build GRCh38
- UniGene ID
- Hs.585648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027004.1 |
|
9 |
1524 |
|
GenBank mRNA |
AK125743.1 |
- |
1 |
162 |
|
BC037859.2 |
- |
9 |
1524 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB054079.1 |
- |
1 |
162 |
|
Chr. 14 - 93905202 - 93909156 [-] |
GenBankmRNA |
AA398784.1 |
- |
1 |
173 |
|
Chr. 14 - 93905277 - 93909164 [-] |
GenBankmRNA |
BI829532.1 |
- |
1 |
133 |
|
Chr. 14 - 93905061 - 93909127 [-] |
GenBankmRNA |
AA446036.1 |
- |
1 |
74 |
|
Chr. 14 - 93905117 - 93909067 [-] |
View Full Product Details