Gene Transcripts

Gene Symbol
PWAR5
Entrez Gene ID
8123
Gene Name
Prader Willi/Angelman region RNA 5
Gene Aliases
D15S226E, PAR-5, PAR5
Chromosome Location
Chr. 15: 24984860 - 24988232 on Build GRCh38
UniGene ID
Hs.660427
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_022008.1
1 1426
GenBank mRNA AF019618.1 - LIT1110
PAR5
1 1236
CN358995.1 -
1 40

View Full Product Details