Gene Transcripts
- Gene Symbol
- PWAR5
- Entrez Gene ID
- 8123
- Gene Name
- Prader Willi/Angelman region RNA 5
- Gene Aliases
- D15S226E, PAR-5, PAR5
- Chromosome Location
- Chr. 15: 24984860 - 24988232 on Build GRCh38
- UniGene ID
- Hs.660427
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_022008.1 |
|
|
1 |
2378 |
|
GenBank mRNA |
AF019618.1 |
- |
LIT1110 PAR5 |
1 |
2188 |
|
View Full Product Details