Gene Transcripts

Gene Symbol
SIAH1
Entrez Gene ID
6477
Gene Name
siah E3 ubiquitin protein ligase 1
Gene Aliases
SIAH1A
Chromosome Location
Chr. 16: 48360531 - 48448440 on Build GRCh38
UniGene ID
Hs.706828
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK023458.1 - 1 993


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA096768.1 - 1 158 Chr. 16 - 48364477 - 48365050 [-]

View Full Product Details