Gene Transcripts

Gene Symbol
RUFY2
Entrez Gene ID
55680
Gene Name
RUN and FYVE domain containing 2
Gene Aliases
RABIP4R, ZFYVE13
Chromosome Location
Chr. 10: 68341107 - 68407302 on Build GRCh38
UniGene ID
Hs.653144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC030097.1 - 1 214


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB502513.1 - 1 208 Chr. 10 - 68355213 - 68355687 [-]
GenBankmRNA BG707400.1 - 1 214 Chr. 10 - 68354953 - 68355693 [-]

View Full Product Details