Gene Transcripts
- Gene Symbol
- MID1
- Entrez Gene ID
- 4281
- Gene Name
- midline 1
- Gene Aliases
- BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY
- Chromosome Location
- Chr. X: 10445310 - 11111264 on Build GRCh38
- UniGene ID
- Hs.27695
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY660579.1 |
- |
4 |
707 |
|
View Full Product Details