Gene Transcripts

Gene Symbol
ST6GALNAC2
Entrez Gene ID
10610
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2
Gene Aliases
SAITL1, SIAT7, SIAT7B, SIATL1, ST6GalNAII, STHM
Chromosome Location
Chr. 17: 76565379 - 76586066 on Build GRCh38
UniGene ID
Hs.592105
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK127567.1 - 1 2653


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU856690.1 - 1 273 Chr. 17 - 76577721 - 76578603 [-]

View Full Product Details