Gene Transcripts

Gene Symbol
SSH2
Entrez Gene ID
85464
Gene Name
slingshot protein phosphatase 2
Gene Aliases
SSH-2, SSH-2L
Chromosome Location
Chr. 17: 29625938 - 29930239 on Build GRCh38
UniGene ID
Hs.654754
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017025253.1 XP_016880742.1 15 1669
GenBank mRNA AF086010.1 - 1 68

View Full Product Details