Gene Transcripts

Gene Symbol
CDRT4
Entrez Gene ID
284040
Gene Name
CMT1A duplicated region transcript 4
Gene Aliases
Chromosome Location
Chr. 17: 15436015 - 15467611 on Build GRCh38
UniGene ID
Hs.164595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK126240.1 - 1 748

View Full Product Details