Gene Transcripts
- Gene Symbol
- ZKSCAN8
- Entrez Gene ID
- 7745
- Gene Name
- zinc finger with KRAB and SCAN domains 8
- Gene Aliases
- LD5-1, ZNF192, ZSCAN40
- Chromosome Location
- Chr. 6: 28141644 - 28159472 on Build GRCh38
- UniGene ID
- Hs.57679
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM727215.1 |
- |
1 |
211 |
|
Chr. 6 - 28158656 - 28159017 [+] |
GenBankmRNA |
BF970561.1 |
- |
|
346 |
|
Chr. - - [-] |
GenBankmRNA |
BX451934.2 |
- |
1 |
336 |
|
Chr. 6 - 28158536 - 28159434 [+] |
GenBankmRNA |
CR606559.1 |
- |
1 |
1077 |
|
Chr. 6 - 28157790 - 28159467 [+] |
GenBankmRNA |
AW958487.1 |
- |
1 |
273 |
|
Chr. 6 - 28158594 - 28159257 [+] |
View Full Product Details