Gene Transcripts
- Gene Symbol
- Gm19345
- Entrez Gene ID
- 100502736
- Gene Name
- predicted gene, 19345
- Gene Aliases
- Chromosome Location
- Chr. 7: 19852003 - 19858355 on Build GRCm38
- UniGene ID
- Mm.445354
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK043814.1 |
- |
1 |
146 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB641623.1 |
- |
1 |
146 |
|
Chr. 7 - 19852003 - 19858039 [+] |
View Full Product Details