Gene Transcripts
- Gene Symbol
- Ccl25
- Entrez Gene ID
- 20300
- Gene Name
- chemokine (C-C motif) ligand 25
- Gene Aliases
- A130072A22Rik, AI852536, CKb15, Scya25, TECK
- Chromosome Location
- Chr. 8: 4325159 - 4360020 on Build GRCm38
- UniGene ID
- Mm.7275
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038019.1 |
- |
1 |
1976 |
|
View Full Product Details