Gene Transcripts
- Gene Symbol
- 4930445E18Rik
- Entrez Gene ID
- 78115
- Gene Name
- RIKEN cDNA 4930445E18 gene
- Gene Aliases
- Chromosome Location
- Chr. 15: 25281439 - 25332648 on Build GRCm38
- UniGene ID
- Mm.160049
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019612.1 |
- |
3 |
179 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY714940.1 |
- |
3 |
179 |
|
Chr. 15 - 25295672 - 25332648 [-] |
View Full Product Details