Gene Transcripts
- Gene Symbol
- 2310057M21Rik
- Entrez Gene ID
- 68277
- Gene Name
- RIKEN cDNA 2310057M21 gene
- Gene Aliases
- 3110040E10Rik, AI429544
- Chromosome Location
- Chr. 7: 131342710 - 131362843 on Build GRCm38
- UniGene ID
- Mm.121122
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014156.1 |
- |
1 |
1263 |
|
AK076241.1 |
- |
1 |
159 |
|
View Full Product Details