Gene Transcripts
- Gene Symbol
- C130046K22Rik
- Entrez Gene ID
- 399609
- Gene Name
- RIKEN cDNA C130046K22 gene
- Gene Aliases
- Chromosome Location
- Chr. 11: 103697724 - 103725573 on Build GRCm38
- UniGene ID
- Mm.351922
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_102388.1 |
|
3 |
1441 |
|
NR_102389.1 |
|
3 |
1430 |
|
GenBank mRNA |
AK142001.1 |
- |
3 |
1425 |
|
View Full Product Details