Gene Transcripts
- Gene Symbol
- 4930588D02Rik
- Entrez Gene ID
- 78226
- Gene Name
- RIKEN cDNA 4930588D02 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.109480
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019831.1 |
- |
1 |
111 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY716010.1 |
- |
1 |
111 |
|
Chr. 15 - 80227939 - 80228414 [-] |
View Full Product Details