Gene Transcripts

Gene Symbol
4930471M09Rik
Entrez Gene ID
75787
Gene Name
RIKEN cDNA 4930471M09 gene
Gene Aliases
4930427C17Rik
Chromosome Location
Chr. 6: 91430460 - 91440854 on Build GRCm38
UniGene ID
Mm.30842
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_045980.1 3 952
GenBank mRNA AK015554.1 - 3 953

View Full Product Details