Gene Transcripts
- Gene Symbol
- 4930485G23Rik
- Entrez Gene ID
- 75808
- Gene Name
- RIKEN cDNA 4930485G23 gene
- Gene Aliases
- 1700038N22Rik
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.401978
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006636.1 |
- |
1 |
557 |
|
View Full Product Details