Gene Transcripts
- Gene Symbol
- 4930590A17Rik
- Entrez Gene ID
- 114677
- Gene Name
- RIKEN cDNA 4930590A17 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.449986
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016377.1 |
- |
4 |
441 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DV658617.1 |
- |
4 |
368 |
|
Chr. 16 - 87909341 - 87952054 [+] |
GenBankmRNA |
BY716024.1 |
- |
|
441 |
|
Chr. - - [-] |
View Full Product Details