Gene Transcripts
- Gene Symbol
- 9430065F17Rik
- Entrez Gene ID
- 77857
- Gene Name
- RIKEN cDNA 9430065F17 gene
- Gene Aliases
- E130110G22Rik
- Chromosome Location
- Chr. 13: 67553170 - 67570904 on Build GRCm38
- UniGene ID
- Mm.160006
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020476.1 |
- |
3 |
172 |
|
AK021377.1 |
- |
3 |
202 |
|
BY126544.1 |
- |
3 |
134 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY721806.1 |
- |
3 |
172 |
|
Chr. 13 - 67553263 - 67564010 [+] |
GenBankmRNA |
BY731297.1 |
- |
3 |
202 |
|
Chr. 13 - 67553234 - 67560809 [+] |
View Full Product Details