Gene Transcripts
- Gene Symbol
- LINC00582
- Entrez Gene ID
- 100287814
- Gene Name
- long intergenic non-protein coding RNA 582
- Gene Aliases
- Chromosome Location
- Chr. 1: 231591292 - 231612090 on Build GRCh38
- UniGene ID
- Hs.25555
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034037.1 |
|
2 |
141 |
|
GenBank mRNA |
AI820788.1 |
- |
2 |
141 |
|
BX092370.1 |
- |
2 |
141 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000448058.1 |
- |
2 |
141 |
|
Chr. 1 - 231591292 - 231612090 [-] |
View Full Product Details