Gene Transcripts
- Gene Symbol
- LINC00589
- Entrez Gene ID
- 619351
- Gene Name
- long intergenic non-protein coding RNA 589
- Gene Aliases
- C8orf75
- Chromosome Location
- Chr. 8: 29721259 - 29748109 on Build GRCh38
- UniGene ID
- Hs.566911
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026765.1 |
|
2 |
186 |
|
GenBank mRNA |
BC003524.1 |
- |
2 |
186 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000506121.3 |
- |
2 |
186 |
|
Chr. 8 - 29721254 - 29748109 [-] |
View Full Product Details