Gene Transcripts
- Gene Symbol
- LINC00645
- Entrez Gene ID
- 100505967
- Gene Name
- long intergenic non-protein coding RNA 645
- Gene Aliases
- Chromosome Location
- Chr. 14: 27612588 - 27639636 on Build GRCh38
- UniGene ID
- Hs.668484
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000555350.1 |
- |
2 |
286 |
|
Chr. 14 - 27634125 - 27636669 [+] |
GENCODE |
ENST00000557359.5 |
- |
4 |
333 |
|
Chr. 14 - 27612588 - 27639636 [+] |
GENCODE |
ENST00000557399.1 |
- |
5 |
361 |
|
Chr. 14 - 27612611 - 27639636 [+] |
View Full Product Details