Gene Transcripts
- Gene Symbol
- AU020206
- Entrez Gene ID
- 100504230
- Gene Name
- expressed sequence AU020206
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.200422
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA210347.1 |
- |
1 |
220 |
|
AK049415.1 |
- |
1 |
2198 |
|
AK080297.1 |
- |
1 |
1210 |
|
View Full Product Details