Gene Transcripts
- Gene Symbol
- LINC01619
- Entrez Gene ID
- 256021
- Gene Name
- long intergenic non-protein coding RNA 1619
- Gene Aliases
- C12orf79
- Chromosome Location
- Chr. 12: 91984976 - 92142831 on Build GRCh38
- UniGene ID
- Hs.651357
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000549802.5 |
- |
5 |
720 |
|
Chr. 12 - 91985090 - 92142671 [-] |
GENCODE |
ENST00000551563.6 |
- |
5 |
589 |
|
Chr. 12 - 91985778 - 92142668 [-] |
GENCODE |
ENST00000552106.5 |
- |
6 |
641 |
|
Chr. 12 - 91985778 - 92142668 [-] |
GENCODE |
ENST00000546975.4 |
- |
5 |
566 |
|
Chr. 12 - 91985778 - 92142668 [-] |
GENCODE |
ENST00000551843.5 |
- |
6 |
642 |
|
Chr. 12 - 91984980 - 92141713 [-] |
View Full Product Details