Gene Transcripts

Gene Symbol
LINC00645
Entrez Gene ID
100505967
Gene Name
long intergenic non-protein coding RNA 645
Gene Aliases
Chromosome Location
Chr. 14: 27612588 - 27639636 on Build GRCh38
UniGene ID
Hs.668484
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_039992.2 5 500
GenBank mRNA BC042069.1 - 2 268
BX538073.1 - 5 477
BX538140.1 - 4 449

View Full Product Details