Gene Transcripts

Gene Symbol
Mcoln3
Entrez Gene ID
171166
Gene Name
mucolipin 3
Gene Aliases
6720490O21Rik, TRPML3, Va
Chromosome Location
Chr. 3: 146093277 - 146141808 on Build GRCm38
UniGene ID
Mm.114683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134160.1 NP_598921.1 10 1283
GenBank mRNA AF475086.1 - 10 1283
AK030819.1 - 10 1403
AK033008.1 - 10 1397
AK035029.1 - 10 1404
AY083531.1 - 10 1281
BC106856.2 - 10 1283 40047039
BC106857.2 - 10 1283 40047042

View Full Product Details