Gene Transcripts

Gene Symbol
Fam149a
Entrez Gene ID
212326
Gene Name
family with sequence similarity 149, member A
Gene Aliases
AW213256, mKIAA4166
Chromosome Location
Chr. 8: 45336715 - 45382291 on Build GRCm38
UniGene ID
Mm.51259
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153535.2 NP_705763.1 7 1963
GenBank mRNA AK034229.1 - 7 1190
AK038552.1 - 7 1178
AK140440.1 - 7 1963
AK141607.1 - 7 1906
AK177317.1 - 1 14
AK212273.1 - 1 14
AK212547.1 - 1 14
AK213568.1 - 1 14
AK218210.1 - 1 14
AK220518.1 - 7 1199
BC035537.1 - 7 1854 5369152

View Full Product Details