Gene Transcripts

Gene Symbol
Olfr290
Entrez Gene ID
258411
Gene Name
olfactory receptor 290
Gene Aliases
MOR254-1
Chromosome Location
Chr. 7: 84915781 - 84916728 on Build GRCm38
UniGene ID
Mm.271340
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146416.2 NP_666528.2 1 766
GenBank mRNA BC127990.1 - 1 770

View Full Product Details