Gene Transcripts
- Gene Symbol
- Olfr1018
- Entrez Gene ID
- 258579
- Gene Name
- olfactory receptor 1018
- Gene Aliases
- MOR260-5
- Chromosome Location
- Chr. 2: 85822973 - 85823908 on Build GRCm38
- UniGene ID
- Mm.223106
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146586.2 |
NP_666797.2
|
1 |
856 |
|
View Full Product Details