Gene Transcripts
- Gene Symbol
- Olfr823
- Entrez Gene ID
- 258668
- Gene Name
- olfactory receptor 823
- Gene Aliases
- MOR210-4
- Chromosome Location
- Chr. 10: 130111841 - 130112788 on Build GRCm38
- UniGene ID
- Mm.377630
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146673.2 |
NP_666884.2
|
1 |
643 |
|
- Gene Symbol
- Olfr824
- Entrez Gene ID
- 258669
- Gene Name
- olfactory receptor 824
- Gene Aliases
- MOR210-3
- Chromosome Location
- Chr. 10: 130126108 - 130127055 on Build GRCm38
- UniGene ID
- Mm.377631
- Species
- Mus musculus
View Full Product Details