Gene Transcripts
- Gene Symbol
- Olfr170
- Entrez Gene ID
- 258959
- Gene Name
- olfactory receptor 170
- Gene Aliases
- MOR273-2
- Chromosome Location
- Chr. 16: 19605725 - 19606666 on Build GRCm38
- UniGene ID
- Mm.377770
- Species
- Mus musculus
- Gene Symbol
- Olfr171
- Entrez Gene ID
- 258960
- Gene Name
- olfactory receptor 171
- Gene Aliases
- MOR273-1, MOR273-5
- Chromosome Location
- Chr. 16: 19624157 - 19625101 on Build GRCm38
- UniGene ID
- Mm.377771
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146958.2 |
NP_667169.2
|
1 |
490 |
|
View Full Product Details