Gene Transcripts

Gene Symbol
Olfr683
Entrez Gene ID
259047
Gene Name
olfactory receptor 683
Gene Aliases
MOR40-1
Chromosome Location
Chr. 7: 105143350 - 105146459 on Build GRCm38
UniGene ID
Mm.223428
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147045.1 NP_667256.1 1 541
GenBank mRNA AF121981.1 - 1 134
BC111830.1 - 1 534
BC148203.1 - 1 541 40055104

View Full Product Details