Gene Transcripts

Gene Symbol
Olfr584
Entrez Gene ID
259056
Gene Name
olfactory receptor 584
Gene Aliases
MOR30-2
Chromosome Location
Chr. 7: 103085520 - 103086479 on Build GRCm38
UniGene ID
Mm.377813
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147054.1 NP_667265.1 1 766
GenBank mRNA BC128007.1 - 1 770

View Full Product Details