Gene Transcripts

Gene Symbol
Gm9758
Entrez Gene ID
381714
Gene Name
predicted gene 9758
Gene Aliases
ENSMUSG00000033219
Chromosome Location
Chr. 5: 14910124 - 14915146 on Build GRCm38
UniGene ID
Mm.368256
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198666.3 NP_941068.1 2 331
GenBank mRNA BC048454.1 - 2 331

View Full Product Details