Gene Transcripts
- Gene Symbol
- Gm9758
- Entrez Gene ID
- 381714
- Gene Name
- predicted gene 9758
- Gene Aliases
- ENSMUSG00000033219
- Chromosome Location
- Chr. 5: 14910124 - 14915146 on Build GRCm38
- UniGene ID
- Mm.368256
- Species
- Mus musculus
- Gene Symbol
- Gm17019
- Entrez Gene ID
- 66773
- Gene Name
- predicted gene 17019
- Gene Aliases
- Chromosome Location
- Chr. 5: 15028950 - 15033527 on Build GRCm38
- UniGene ID
- Mm.368256
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_182957.1 |
NP_892002.1
|
5 |
1012 |
|
View Full Product Details