Gene Transcripts
- Gene Symbol
- Olfr242
- Entrez Gene ID
- 406175
- Gene Name
- olfactory receptor 242
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.246532
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010974.1 |
NP_035104.1
|
|
570 |
|
GenBank mRNA |
X89693.1 |
- |
|
196 |
|
- Gene Symbol
- Olfr27
- Entrez Gene ID
- 258826
- Gene Name
- olfactory receptor 27
- Gene Aliases
- MOR171-6, MTPCR56
- Chromosome Location
- Chr. 9: 39128167 - 39145072 on Build GRCm38
- UniGene ID
- Mm.246532
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146829.2 |
NP_667040.2
|
2 |
640 |
|
View Full Product Details