Gene Transcripts

Gene Symbol
Aldh3b2
Entrez Gene ID
621603
Gene Name
aldehyde dehydrogenase 3 family, member B2
Gene Aliases
AI848594, C130048D07Rik
Chromosome Location
Chr. 19: 3972328 - 3981665 on Build GRCm38
UniGene ID
Mm.34137
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177438.1 NP_001170909.1 10 1637
GenBank mRNA AK048310.1 - 498
AK136505.1 - 10 1641
BC100370.1 - 495

View Full Product Details