Gene Transcripts
- Gene Symbol
- Gm7894
- Entrez Gene ID
- 666023
- Gene Name
- predicted gene 7894
- Gene Aliases
- EG666023
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.396234
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK131747.1 |
- |
2 |
147 |
|
View Full Product Details