Gene Transcripts
- Gene Symbol
- Foxg1
- Entrez Gene ID
- 24370
- Gene Name
- forkhead box G1
- Gene Aliases
- BF1A, Fkhl1, RATBF1A
- Chromosome Location
- Chr. 6: 69971227 - 69974037 on Build Rnor_6.0
- UniGene ID
- Rn.9864
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012560.2 |
NP_036692.1
|
1 |
1387 |
|
GenBank mRNA |
M87634.1 |
- |
1 |
1388 |
|
View Full Product Details