Gene Transcripts
- Gene Symbol
- Pdlim4
- Entrez Gene ID
- 24915
- Gene Name
- PDZ and LIM domain 4
- Gene Aliases
- H-Rev18, Ril
- Chromosome Location
- Chr. 10: 39390578 - 39405311 on Build Rnor_6.0
- UniGene ID
- Rn.34221
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017062.2 |
NP_058758.1
|
1, 2 |
170 |
|
GenBank mRNA |
X76454.1 |
- |
1, 2 |
171 |
|
View Full Product Details