Gene Transcripts

Gene Symbol
Dlg1
Entrez Gene ID
25252
Gene Name
discs, large homolog 1 (Drosophila)
Gene Aliases
Dlgh1, SAP97
Chromosome Location
Chr. 11: 72164566 - 72378895 on Build Rnor_6.0
UniGene ID
Rn.89331
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012788.1 NP_036920.1 10 1358
GenBank mRNA FQ211533.1 - 390
U14950.1 - 10 1358

View Full Product Details