Gene Transcripts
- Gene Symbol
- Sstr4
- Entrez Gene ID
- 25555
- Gene Name
- somatostatin receptor 4
- Gene Aliases
- Smstr4
- Chromosome Location
- Chr. 3: 142739781 - 142741644 on Build Rnor_6.0
- UniGene ID
- Rn.9936
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013036.2 |
NP_037168.1
|
1 |
736 |
|
GenBank mRNA |
U04738.1 |
- |
1 |
736 |
|
View Full Product Details