Gene Transcripts

Gene Symbol
Ywhah
Entrez Gene ID
25576
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta
Gene Aliases
14-3-3e
Chromosome Location
Chr. 14: 83053041 - 83062424 on Build Rnor_6.0
UniGene ID
Rn.3324
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013052.1 NP_037184.1 2 289
GenBank mRNA BC081825.1 - 2 314
D17445.1 - 2 289

View Full Product Details