Gene Transcripts
- Gene Symbol
- Csn3
- Entrez Gene ID
- 29188
- Gene Name
- casein kappa
- Gene Aliases
- Csn10, Csnk
- Chromosome Location
- Chr. 14: 21803738 - 21810291 on Build Rnor_6.0
- UniGene ID
- Rn.10746
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031562.1 |
NP_113750.1
|
2 |
94 |
|
GenBank mRNA |
K02598.1 |
- |
2 |
94 |
|
View Full Product Details