Gene Transcripts
- Gene Symbol
- Cit
- Entrez Gene ID
- 83620
- Gene Name
- citron rho-interacting serine/threonine kinase
- Gene Aliases
- Chromosome Location
- Chr. 12: 46334669 - 46494155 on Build Rnor_6.0
- UniGene ID
- Rn.89069
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001029911.1 |
NP_001025082.1
|
23, 24 |
3066 |
|
XM_006249469.2 |
XP_006249531.1
|
24, 25 |
3044 |
|
XM_006249470.2 |
XP_006249532.1
|
24, 25 |
3044 |
|
XM_006249471.2 |
XP_006249533.1
|
24, 25 |
3044 |
|
XM_006249472.2 |
XP_006249534.1
|
24, 25 |
3044 |
|
XM_006249477.2 |
XP_006249539.1
|
14, 15 |
2232 |
|
XM_008769269.1 |
XP_008767491.1
|
24, 25 |
3044 |
|
XM_008769270.1 |
XP_008767492.1
|
24, 25 |
3000 |
|
XM_008769271.1 |
XP_008767493.1
|
23, 24 |
3070 |
|
GenBank mRNA |
AF039218.1 |
- |
14, 15 |
2170 |
|
View Full Product Details