Gene Transcripts

Gene Symbol
Fstl3
Entrez Gene ID
114031
Gene Name
follistatin-like 3 (secreted glycoprotein)
Gene Aliases
Flrg
Chromosome Location
Chr. 7: 12806031 - 12810556 on Build Rnor_6.0
UniGene ID
Rn.48835
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053629.3 NP_446081.1 2 241
GenBank mRNA AB021295.1 - 2 241
AB071603.1 - 262

View Full Product Details