Gene Transcripts

Gene Symbol
Psmd9
Entrez Gene ID
161475
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 9
Gene Aliases
Bridge, Bridge-1
Chromosome Location
Chr. 12: 38786539 - 38805121 on Build Rnor_6.0
UniGene ID
Rn.24127
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130430.1 NP_569114.1 2 693
GenBank mRNA AF067728.1 - 2 693
FQ214178.1 - 237
FQ225583.1 - 226
FQ227319.1 - 226
FQ229304.1 - 259

Gene Symbol
LOC103694906
Entrez Gene ID
103694906
Gene Name
26S proteasome non-ATPase regulatory subunit 9-like
Gene Aliases
Chromosome Location
Chr. Un|NW_007906959: 139 - 7101 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773903.1 XP_008772125.1 1 430

View Full Product Details