Gene Transcripts

Gene Symbol
Oas1b
Entrez Gene ID
246268
Gene Name
2-5 oligoadenylate synthetase 1B
Gene Aliases
Chromosome Location
Chr. 12: 41255762 - 41266430 on Build Rnor_6.0
UniGene ID
Rn.14530
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144752.2 NP_653353.2 1 80
GenBank mRNA AF068268.1 - 92
BC091121.1 - 1 80 7383389

View Full Product Details